ContactUsLinkComponent
Precision density hydrometer, range: 1.800-2.000 in 0.002 G/Cm3 total length: 280 mm without thermometer adjusting temperature: 20c , suitable for official certification 1 * 1 items
Catalog # SCNL3071007
Precision density hydrometer, range: 1.800-2.000 in 0.002 G/Cm3 total length: 280 mm without thermometer adjusting temperature: 20c , suitable for official certification 1 * 1 items
Catalog # SCNL3071007
Frequently Bought Together