ContactUsLinkComponent
Heating output, reinforced - 400 V option model, For faster heating up time and improved recovery time max. 300 C not in conjunction with load management option 1 * 1 items
Catalog # BIND8012-1061
Supplier:  Binder
Heating output, reinforced - 400 V option model, For faster heating up time and improved recovery time max. 300 C not in conjunction with load management option 1 * 1 items
Catalog # BIND8012-1061
Supplier:  Binder
Frequently Bought Together