ContactUsLinkComponent
Anschütztherm., +198+252:0,1°C, solid stem,white backed, gallium filled, 340x5-6mm, total immersion, with button at top, government calibrated, with DKD-Certificate (DAkks) with 3 test points 1 * 1 items
Catalog # AMARN60080-GA-DKD3
Supplier:  Amarell
Anschütztherm., +198+252:0,1°C, solid stem,white backed, gallium filled, 340x5-6mm, total immersion, with button at top, government calibrated, with DKD-Certificate (DAkks) with 3 test points 1 * 1 items
Catalog # AMARN60080-GA-DKD3
Supplier:  Amarell
Frequently Bought Together