ContactUsLinkComponent
Pipets, unserialized reusable to deliver volumetric class a , capacity (ml)-25, tolerance(ml)± 0.03, overall length (mm)-505, color of coding band-blue. 1 * 6 items
Catalog # SCERSP37002-5
Supplier:  DWK Life Sciences
Pipets, unserialized reusable to deliver volumetric class a , capacity (ml)-25, tolerance(ml)± 0.03, overall length (mm)-505, color of coding band-blue. 1 * 6 items
Catalog # SCERSP37002-5
Supplier:  DWK Life Sciences
Frequently Bought Together