Platform with 16/8/8 sockets for 1.5/0.5/0.2ml microtest tubes 1 * 1 items
GRANSV-16/8
Platform with 16/8/8 sockets for 1.5/0.5/0.2ml microtest tubes 1 * 1 items
GRANSV-16/8
Frequently Bought Together
DISPERSING AGGREGATE PT-DA 20/2MEC-F193 1 * 1 items
: KINEMATICA
Magnetic Stirrers, RT basic
: Thermo Fisher Scientific
Laboratory mixers
: WARING BLENDOR
PLATFORM FOR 10 X 10 ML (12 MM) TUBES 1 * 1 items
: GRANT INSTRUMENTS
Condenser, air, CondenSyn™
: ASYNT