ContactUsLinkComponent
Florisil® for Hydrocarbon Index analysis, 60-100 mesh For Hydrocarbon Index determination according to EN ISO 9377-2 (H53). Empirical formula MgO · 3,6 SiO2 · 1,53 OH Density (D) 2,51 CAS No.[1343-88-0] EG-Nr. 215-681-1 1 * 500 g
Catalog # ROTHCN39.3
Supplier:  Roth Carl
Florisil® for Hydrocarbon Index analysis, 60-100 mesh For Hydrocarbon Index determination according to EN ISO 9377-2 (H53). Empirical formula MgO · 3,6 SiO2 · 1,53 OH Density (D) 2,51 CAS No.[1343-88-0] EG-Nr. 215-681-1 1 * 500 g
Catalog # ROTHCN39.3
Supplier:  Roth Carl
Frequently Bought Together