ContactUsLinkComponent
Thermom. similar to ASTM 110 C, solid stem, white backed, 133,6+136,4:0 ,05C, blue spezial filled, total immersion, 305x6-8mm, suitable for government calibration, durable pigment with ice point scale 1 * 1 items
Catalog # AMARA301474-CB
Supplier:  Amarell
Thermom. similar to ASTM 110 C, solid stem, white backed, 133,6+136,4:0 ,05C, blue spezial filled, total immersion, 305x6-8mm, suitable for government calibration, durable pigment with ice point scale 1 * 1 items
Catalog # AMARA301474-CB
Supplier:  Amarell
Frequently Bought Together