Microscope cover glasses, 29,9x29,9 mm, borosilicate glass D 263 M ,thickness 0.7 mm +/- 0.05 mm, 25 pcs./cardboard box, 250 pcs./carton, MARIENFELD SUPERIOR 1 * 10.000 items
Catalog # MARI0109999094
Supplier: MARIENFELD
Frequently Bought Together