0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SMALL DISPOSABLE POLYPROPYLENE WEIGHING FUNNEL. FITS INTO 5-50ML VOLUMETRIC FLASKS. 50/PACK, 1 * 50 items
Catalog # TRADDPWFPP1S
Supplier:
TRADEWINDS DIRECT
Expand image
SMALL DISPOSABLE POLYPROPYLENE WEIGHING FUNNEL. FITS INTO 5-50ML VOLUMETRIC FLASKS. 50/PACK, 1 * 50 items
Catalog # TRADDPWFPP1S
Supplier:
TRADEWINDS DIRECT
Frequently Bought Together
VWR® Nitrile Light Long, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Latex Gloves
Supplier: VWR Collection
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Laboratory Markers
Supplier: VWR Collection
Lead-free steam indicator tapes, Comply™
Supplier: 3M MEDICAL
VWR®, Nitrile Gloves
Supplier: VWR Collection