EDC is a heterobifunctional, water-soluble, zero-length carbodiimide crosslinker that is used to couple carboxyl groups to primary a mines. EDC activates carboxyl groups first and forms amine reactive O-acylisourea imtermediate that spontaneously rea 1 * 25 g
Catalog # GENOBC25-25
Supplier: G-Biosciences