A Xaf1 (XIAP-associated factor 1) ELISA with a senstivity of 46.9pg/ml and a range of 78-5000pg/ml. Uses the detection method Sandwi ch ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8261
Supplier: G-Biosciences