ContactUsLinkComponent
A Mecp2 (Methyl-CpG-binding protein 2) ELISA with a senstivity of 46.875pg/ml and a range of 78.125-5000pg/ml. Uses the detection me thod Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8291
Supplier:  G-Biosciences
A Mecp2 (Methyl-CpG-binding protein 2) ELISA with a senstivity of 46.875pg/ml and a range of 78.125-5000pg/ml. Uses the detection me thod Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8291
Supplier:  G-Biosciences