A Aldoc (Fructose-bisphosphate aldolase C) ELISA with a senstivity of 46.9pg/ml and a range of 78-5000pg/ml. Uses the detection meth od Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8280
Supplier: G-Biosciences