ContactUsLinkComponent
An ELISA kit for the detection of Visfatin C termina (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.2 81ng/ml. 1 * 1 KIT
Catalog # GENOIT4575
Supplier:  G-Biosciences
An ELISA kit for the detection of Visfatin C termina (Human). This uses Sandwich ELISA, Double Antibody and has a sensitivity of 0.2 81ng/ml. 1 * 1 KIT
Catalog # GENOIT4575
Supplier:  G-Biosciences