ContactUsLinkComponent
An ELISA kit for the detection of Testosterone (Human). This uses Competitive ELISA, Coated with Antigen and has a sensitivity of 0. 188ng/ml. 1 * 1 KIT
Catalog # GENOIT4768
Supplier:  G-Biosciences
An ELISA kit for the detection of Testosterone (Human). This uses Competitive ELISA, Coated with Antigen and has a sensitivity of 0. 188ng/ml. 1 * 1 KIT
Catalog # GENOIT4768
Supplier:  G-Biosciences
Frequently Bought Together