JAW MES Buffered Saline Packs are ready to use dry-blend powder pouches designed to make MES buffer saline which is amine and carbo xyl-free and used for carbodiimide (EDC) crosslinking and conjugation reactions. 1 * 10 items
Catalog # GENO786-1208
Supplier: G-Biosciences