ContactUsLinkComponent
Conventional stain used for H&E staining as well as counterstaining in immunohistochemistry (IHC) and cytology procedures. Ready to use formulation optimized for progressive nuclear staining with lower risk of overstaining than other formulations. 1 * 4 L
Catalog # GENO786-1049
Supplier:  G-Biosciences
Conventional stain used for H&E staining as well as counterstaining in immunohistochemistry (IHC) and cytology procedures. Ready to use formulation optimized for progressive nuclear staining with lower risk of overstaining than other formulations. 1 * 4 L
Catalog # GENO786-1049
Supplier:  G-Biosciences