ContactUsLinkComponent
G-Biosciences immobilized antibiotics are prepared by coupling antibiotic to preactivated 6% cross-linked agarose. The covalent coup ling to agarose (resin) is achieved by cross-linking amino or carboxyl group depending upon the antibiotic that is co 1 * 10 mL
Catalog # GENO786-1344
Supplier:  G-Biosciences
G-Biosciences immobilized antibiotics are prepared by coupling antibiotic to preactivated 6% cross-linked agarose. The covalent coup ling to agarose (resin) is achieved by cross-linking amino or carboxyl group depending upon the antibiotic that is co 1 * 10 mL
Catalog # GENO786-1344
Supplier:  G-Biosciences