ContactUsLinkComponent
FCCP [Carbonyl cyanide 4-(trifluoromethoxy) phenylhydrazone] is a potent uncoupler of oxidative phosphorylation in mitochondria. It is used as a positive control for mitochondrial membrane depolarization. FCCP disrupts the synthesis of ATP by trnas 1 * 10 mg
Catalog # GENO786-1316
Supplier:  G-Biosciences
FCCP [Carbonyl cyanide 4-(trifluoromethoxy) phenylhydrazone] is a potent uncoupler of oxidative phosphorylation in mitochondria. It is used as a positive control for mitochondrial membrane depolarization. FCCP disrupts the synthesis of ATP by trnas 1 * 10 mg
Catalog # GENO786-1316
Supplier:  G-Biosciences
Frequently Bought Together