ContactUsLinkComponent
Luciferase (Firefly, Recombinant) is recombinant 62 kDa protein expressed in E. Coli. Single band purity, >99 % by SDS-PAGE. High s pecific activity: ≥1 x 10E10 units/mg 1 * 100 µG
Catalog # GENO786-1308
Supplier:  G-Biosciences
Luciferase (Firefly, Recombinant) is recombinant 62 kDa protein expressed in E. Coli. Single band purity, >99 % by SDS-PAGE. High s pecific activity: ≥1 x 10E10 units/mg 1 * 100 µG
Catalog # GENO786-1308
Supplier:  G-Biosciences
Frequently Bought Together