Diethyl ether EMPROVE ESSENTIAL Ph Eur,BP 1 * 190 L
Catalog # 1.00926.6190
Supplier: MERCK PRODUCTION CHEMICALS
Diethyl ether EMPROVE ESSENTIAL Ph Eur,BP 1 * 190 L
Catalog # 1.00926.6190
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Disodium hydrogen phosphate dodecahydrate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
2-Ethoxyethanol ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals