GENESYS 140 Visible Spectrophotometer with UK Power Cable Visible range split/dual beam scanning spectrophotometer Supports single sample measurements Supports optional 8-position rotary cell changer 325nm - 1100nm -3.5A to +3.5A photometric acc 1 * 1 items
Catalog # 634-1112
Supplier: Thermo Fisher Scientific
Frequently Bought Together