ContactUsLinkComponent
Single Cell Cuvette Holder + base for GENESYS 100 Series Holds one 10mm pathlength standard square cuvette Also supports shorter p athlength cells with an appropriate spacer Includes magnetic base plate. 1 * 1 items
Catalog # 634-1093
Single Cell Cuvette Holder + base for GENESYS 100 Series Holds one 10mm pathlength standard square cuvette Also supports shorter p athlength cells with an appropriate spacer Includes magnetic base plate. 1 * 1 items
Catalog # 634-1093
Frequently Bought Together