ContactUsLinkComponent
Sipper Pump For GENESYS with UK plug External pump, power supply and cable Controlled by instrument software, Includes 30 cm #16 silicone tubing, Requires Accessory Kit For Hose Entry/Exit for use with GEN 40 or 50, Requires Sipper Insert Accessory 1 * 1 items
Catalog # 634-1092
Sipper Pump For GENESYS with UK plug External pump, power supply and cable Controlled by instrument software, Includes 30 cm #16 silicone tubing, Requires Accessory Kit For Hose Entry/Exit for use with GEN 40 or 50, Requires Sipper Insert Accessory 1 * 1 items
Catalog # 634-1092
Frequently Bought Together