Peltier Thermostatted Single Cell Holder for GENESYS 100 and BioMate 160 w/ UK plug. Air-cooled Peltier system. Fits inside the sam ple compartment of the instrument. Cell holder for 10mm pathlength square cells. Temperature range 20C-60C. Magne va 1 * 1 items
Catalog # 634-1107
Supplier: Thermo Fisher Scientific
Frequently Bought Together