ContactUsLinkComponent
Gdf15 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat GDF-15 in cell culture super nates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5189
Supplier:  Abnova
Gdf15 (Rat) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of rat GDF-15 in cell culture super nates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5189
Supplier:  Abnova