£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Pestizide - Mix 5, je 10 ìg/ml in Methanol, 5 ml Triethylphosphat 78-40-0 Chlorpyriphos-ethyl 2921-88-2 Fenitrothion 122-14-5 Triphenylphosphinoxid 791-28-6 Trifluralin 1582-09-8 1 * 5 mL
PROOSL24733
:
LGC Standards PROMOCHEM
Pestizide - Mix 5, je 10 ìg/ml in Methanol, 5 ml Triethylphosphat 78-40-0 Chlorpyriphos-ethyl 2921-88-2 Fenitrothion 122-14-5 Triphenylphosphinoxid 791-28-6 Trifluralin 1582-09-8 1 * 5 mL
PROOSL24733
:
LGC Standards PROMOCHEM
Frequently Bought Together
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
pH indicator strips, MQuant®, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals