ContactUsLinkComponent
custom-made product, three neck round bottom flask, 500ml, center neck NS 29/32, side necks NS 19/26, amber 1 * 1 items
Catalog # LENZ81004023
custom-made product, three neck round bottom flask, 500ml, center neck NS 29/32, side necks NS 19/26, amber 1 * 1 items
Catalog # LENZ81004023
Frequently Bought Together