HWS-Dreihalsaufsatz, aus Duran mit parallelen Hälsen, 1 x NSK 29/32, 3 x NSH 29/32 1 * 1 items
HWSA2703030
HWS-Dreihalsaufsatz, aus Duran mit parallelen Hälsen, 1 x NSK 29/32, 3 x NSH 29/32 1 * 1 items
HWSA2703030
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection