ContactUsLinkComponent
Analog output 4-20 mA - for temperature and humidity values (output not adjustable) 1 * 1 items
Catalog # 390-1202
Supplier:  Binder
Analog output 4-20 mA - for temperature and humidity values (output not adjustable) 1 * 1 items
Catalog # 390-1202
Supplier:  Binder
Frequently Bought Together