ContactUsLinkComponent
4-ft (1.2m) Hydraulic Lift Base Stand for use with Purifier Class II Biosafety Cabinets. Supports loads to 1300 lbs. Powder-coated steel. Height adjustable: 17.2 (43.7cm) to 36.8 (93.5cm) work surface height. Casters. 230V, UK plug.EA 1 * 1 items
Catalog # LBCP3780331
Supplier:  LABCONCO
4-ft (1.2m) Hydraulic Lift Base Stand for use with Purifier Class II Biosafety Cabinets. Supports loads to 1300 lbs. Powder-coated steel. Height adjustable: 17.2 (43.7cm) to 36.8 (93.5cm) work surface height. Casters. 230V, UK plug.EA 1 * 1 items
Catalog # LBCP3780331
Supplier:  LABCONCO
Frequently Bought Together