ContactUsLinkComponent
Reactor Lid, PTFE. Suitable for glass reactor DN 100 with SCHOTT® flat flange. With screw in threads for connection of transition fittings and a center neck NPT 1/2” 1 * 1 items
Catalog # BOHLB150-12
Supplier:  Bohlender
undefined
Reactor Lid, PTFE. Suitable for glass reactor DN 100 with SCHOTT® flat flange. With screw in threads for connection of transition fittings and a center neck NPT 1/2” 1 * 1 items
Catalog # BOHLB150-12
Supplier:  Bohlender
Frequently Bought Together