ContactUsLinkComponent
Electronic indoor-/outdoor thermometer Dual Thermo Max/min , -50...+70:0,1C, with temperature alarm, max/min function, PVC cable for outdoor-measurements c. 3m, dimensions 65x67x23mm 1 * 1 items
Catalog # AMARE906800
Supplier:  Amarell
Electronic indoor-/outdoor thermometer Dual Thermo Max/min , -50...+70:0,1C, with temperature alarm, max/min function, PVC cable for outdoor-measurements c. 3m, dimensions 65x67x23mm 1 * 1 items
Catalog # AMARE906800
Supplier:  Amarell
Frequently Bought Together