ContactUsLinkComponent
Hydrometer DIN 12794/BS 718, L50, 0,90-0,95:0,0005g/cm3, ref Temp 20C, without therm, max 335mm long, government calibrated, with DKD-Certificate with 3 test points 1 * 1 items
  AMARH806212/DKD3
 :  Amarell
Hydrometer DIN 12794/BS 718, L50, 0,90-0,95:0,0005g/cm3, ref Temp 20C, without therm, max 335mm long, government calibrated, with DKD-Certificate with 3 test points 1 * 1 items
  AMARH806212/DKD3
 :  Amarell
Frequently Bought Together