ContactUsLinkComponent
SONOREX DIGITEC DT 514 BH-RC, ultrasonic bath, 230 V~(± 10%) 50/60 Hz, ID 325x300x200 mm 18,7 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15,30 min and continuous, ultrasonic peak output 860 W, HF output 215 Weff., heater 20 - 80 °C (68-176°F) 1 * 1 items
Catalog # BAND3095-GB
SONOREX DIGITEC DT 514 BH-RC, ultrasonic bath, 230 V~(± 10%) 50/60 Hz, ID 325x300x200 mm 18,7 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15,30 min and continuous, ultrasonic peak output 860 W, HF output 215 Weff., heater 20 - 80 °C (68-176°F) 1 * 1 items
Catalog # BAND3095-GB
Frequently Bought Together