Blasticidin S hydrochloride, 10 mg/ml in HEPES buffer, sterile-filtered 1 * 5 items
Catalog # J67216.8X
Supplier: Thermo Fisher Scientific
Blasticidin S hydrochloride, 10 mg/ml in HEPES buffer, sterile-filtered 1 * 5 items
Catalog # J67216.8X
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Ethyl acetate 99.5+% for spectroscopy
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Nile red 99%, pure
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butanol NF
Supplier: Spectrum Chemical
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
2-Propanol 99.5%, gradient grade for HPLC
Supplier: Thermo Fisher Scientific