ContactUsLinkComponent
Glove Box Vitrum with Transfer Chamber. Aluminium frame with panels made of borosilicate glass, front panel with two glove ports, right side panel and bottom side base panel made of aluminium, top side of base panel made of stainless steel. Including 1 * 1 items
Catalog # BOHLV1986-08
Supplier:  Bohlender
undefined
Glove Box Vitrum with Transfer Chamber. Aluminium frame with panels made of borosilicate glass, front panel with two glove ports, right side panel and bottom side base panel made of aluminium, top side of base panel made of stainless steel. Including 1 * 1 items
Catalog # BOHLV1986-08
Supplier:  Bohlender
Frequently Bought Together