ContactUsLinkComponent
VISOCOLOR ECO Detergents anionic test kit measuring range: 0.1-5.0 mg/L MBAS sufficient for 50 determinations __UN 3316 Chemical kit 9 II 0.100 L ADR/GGVSEB M11 (E) ADR 3.3.1/251: LQ PAX 960 10 Kg CAO 960 10 Kg 1 * 1 items
Catalog # MANA931050
Supplier:  MACHEREY-NAGEL
undefined
VISOCOLOR ECO Detergents anionic test kit measuring range: 0.1-5.0 mg/L MBAS sufficient for 50 determinations __UN 3316 Chemical kit 9 II 0.100 L ADR/GGVSEB M11 (E) ADR 3.3.1/251: LQ PAX 960 10 Kg CAO 960 10 Kg 1 * 1 items
Catalog # MANA931050
Supplier:  MACHEREY-NAGEL