ContactUsLinkComponent
Durapore multilayer Optiscale - 0.45 / 0.22 ìm - Multilayer PVDF hydrophilic membrane - 47 mm diameter / 13.8 cm² - 6 mm Hose Barb Inlet and Outlet -3 per package 1 * 1 items
Catalog # MILXSHGLA47HH3
Supplier:  Merck
Durapore multilayer Optiscale - 0.45 / 0.22 ìm - Multilayer PVDF hydrophilic membrane - 47 mm diameter / 13.8 cm² - 6 mm Hose Barb Inlet and Outlet -3 per package 1 * 1 items
Catalog # MILXSHGLA47HH3
Supplier:  Merck
Frequently Bought Together