ContactUsLinkComponent
This funnel for long and bulk solids can be used to process most samples. This funnel can be used to process free flowing materials, as well as to feed straw, film and other materials in their full length. 1 * 1 items
Catalog # FRIT15.1550.00
Supplier:  FRITSCH
This funnel for long and bulk solids can be used to process most samples. This funnel can be used to process free flowing materials, as well as to feed straw, film and other materials in their full length. 1 * 1 items
Catalog # FRIT15.1550.00
Supplier:  FRITSCH
Frequently Bought Together