ContactUsLinkComponent
Grinding bowl tempered steel, with steel casing, 45 ml volume for PULVERISETTE 7 premium line null
Catalog # FRIT50.9677.00
Supplier:  FRITSCH
Grinding bowl tempered steel, with steel casing, 45 ml volume for PULVERISETTE 7 premium line null
Catalog # FRIT50.9677.00
Supplier:  FRITSCH
Frequently Bought Together