ContactUsLinkComponent
For comminution of dry sample material with soft to medium-hard consistency, for fibrous materials or cellulosic materials as well as for sample preparation according to RoHS. null
Catalog # FRIT15.4050.00
Supplier:  FRITSCH
For comminution of dry sample material with soft to medium-hard consistency, for fibrous materials or cellulosic materials as well as for sample preparation according to RoHS. null
Catalog # FRIT15.4050.00
Supplier:  FRITSCH
Frequently Bought Together