ContactUsLinkComponent
Accessory kit for increased chemical resistance consisting of seals, O-rings, flow plate and hoses for small volume wet dispersion u nit SVA article no. 22.8800.00 for Laser Particle Sizers ANALYSETTE 22 MicroTec plus and NanoTec plus 1 * 1 items
Catalog # FRIT22.8855.00
Supplier:  FRITSCH
Accessory kit for increased chemical resistance consisting of seals, O-rings, flow plate and hoses for small volume wet dispersion u nit SVA article no. 22.8800.00 for Laser Particle Sizers ANALYSETTE 22 MicroTec plus and NanoTec plus 1 * 1 items
Catalog # FRIT22.8855.00
Supplier:  FRITSCH
Frequently Bought Together