ContactUsLinkComponent
The bottom sieves are made of stainless steel trapezoidal resp. round perforation sheets metal. Larger openings mean faster through put, less time in the mill and less wear. null
Catalog # FRIT16.6715.10
Supplier:  FRITSCH
The bottom sieves are made of stainless steel trapezoidal resp. round perforation sheets metal. Larger openings mean faster through put, less time in the mill and less wear. null
Catalog # FRIT16.6715.10
Supplier:  FRITSCH
Frequently Bought Together