ContactUsLinkComponent
test sieve 400/65 mm mesh = 1/2“ = 12.5 mm - ASTM - E-11-1995 - mesh 1 * 1 items
Catalog # FRIT34.1010.02
Supplier:  FRITSCH
test sieve 400/65 mm mesh = 1/2“ = 12.5 mm - ASTM - E-11-1995 - mesh 1 * 1 items
Catalog # FRIT34.1010.02
Supplier:  FRITSCH
Frequently Bought Together