ContactUsLinkComponent
Human APOC3 full-length ORF (AAH27977.1, 1 a.a. - 99 a.a.) recombinant protein with GST tag at N-terminal. Application: AP,Array,EL ISA,WB-Re 1 * 10 µG
Catalog # ABNOH00000345-P01S
Supplier:  Abnova
Human APOC3 full-length ORF (AAH27977.1, 1 a.a. - 99 a.a.) recombinant protein with GST tag at N-terminal. Application: AP,Array,EL ISA,WB-Re 1 * 10 µG
Catalog # ABNOH00000345-P01S
Supplier:  Abnova