BUFFER B CX-1 PH 10.2 , CX-1 PH GRADIENT BUFFER B (PH 10.2) 500ML 1 * 500 mL
Catalog # 555-0673
Supplier: Thermo Fisher Scientific
BUFFER B CX-1 PH 10.2 , CX-1 PH GRADIENT BUFFER B (PH 10.2) 500ML 1 * 500 mL
Catalog # 555-0673
Supplier: Thermo Fisher Scientific
Frequently Bought Together
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
VWR®, OCT Embedding matrix for frozen sections
Supplier: VWR Chemicals
QC KIT DILUENT SOLUTION 1 * 500 mL
Supplier: Tecan
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, Fluka™
Supplier: Honeywell Chemicals