ContactUsLinkComponent
10ml PrecisionULTRA qPCR Master Mix for capillary LightCycler 1.0-2.0 and inert blue dye (10x 1ml) 1 * 1.000 Tests
Catalog # PULTRABLUECL10
Supplier:  PRIMERDESIGN
10ml PrecisionULTRA qPCR Master Mix for capillary LightCycler 1.0-2.0 and inert blue dye (10x 1ml) 1 * 1.000 Tests
Catalog # PULTRABLUECL10
Supplier:  PRIMERDESIGN