ContactUsLinkComponent
ASTM-Thermom., S63C, white backed, -8+32:0,1C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.411x7-8mm, durable pigment, government calibrated with DKD Certificate with 5 test points 1 * 1 items
Catalog # AMARA300860-CBDKD5
Supplier:  Amarell
ASTM-Thermom., S63C, white backed, -8+32:0,1C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.411x7-8mm, durable pigment, government calibrated with DKD Certificate with 5 test points 1 * 1 items
Catalog # AMARA300860-CBDKD5
Supplier:  Amarell
Frequently Bought Together