ContactUsLinkComponent
Test Tube Cell Holder for Novaspec III+ and Novaspec Pro. Spring loaded and compatible with test tubes that are 10 mm to 18 mm in diameter. 1 * 1 items
Catalog # WAPR80-3007-12
Supplier:  Biochrom
Test Tube Cell Holder for Novaspec III+ and Novaspec Pro. Spring loaded and compatible with test tubes that are 10 mm to 18 mm in diameter. 1 * 1 items
Catalog # WAPR80-3007-12
Supplier:  Biochrom
Frequently Bought Together